Learn More
Biomatik Corporation IL-9 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51860
Encompass_Preferred
IL-9 (Human) Sandwich ELISA Kit for detecting IL-9 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 50-3000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P15248. Gene ID: 3578. Target Synonyms: Interleukin-9; Cytokine P40; T-cell growth factor P40. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1832