Learn More
Biomatik Corporation MIG (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51867
Encompass_Preferred
MIG (Human) Sandwich ELISA Kit for detecting MIG in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 16-1000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q07325. Gene ID: 4283. Target Synonyms: C-X-C motif chemokine 9; Gamma-interferon-induced monokine; Monokine induced by interferon-gamma; SCYB9; CMK. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1836