Learn More
Biomatik Corporation NAP-2 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51869
Encompass_Preferred
NAP-2 (Human) Sandwich ELISA Kit for detecting NAP-2 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 16-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P02775. Gene ID: 5473. Target Synonyms: CXCL7; Neutrophil-Activating Peptide 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1838