Learn More
Biomatik Corporation Resistin (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51875
Encompass_Preferred
Resistin (Human) Sandwich ELISA Kit for detecting Resistin in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 16-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9HD89. Gene ID: 56729. Target Synonyms: Adipose tissue-specific secretory factor; ADSF; C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein; Cysteine-rich secreted protein A12-alpha-like 2; Cysteine-rich secreted protein FIZZ3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1843