Learn More
Biomatik Corporation SCF (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51877
Encompass_Preferred
SCF (Human) Sandwich ELISA Kit for detecting SCF in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 16-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P21583. Gene ID: 4254. Target Synonyms: Kit ligand; Mast cell growth factor; Stem cell factor; c-Kit ligand; MGF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1845