Learn More
Biomatik Corporation TACI (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51879
Encompass_Preferred
TACI (Human) Sandwich ELISA Kit for detecting TACI in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 16-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O14836. Gene ID: 23495. Target Synonyms: Tumor necrosis factor receptor superfamily member 13B; Transmembrane activator and CAML interactor; CD267. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1847