Learn More
Biomatik Corporation EGF (Murine) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51888
Encompass_Preferred
EGF (Murine) Sandwich ELISA Kit for detecting EGF in plasma, serum, lysate. Species Reactivity: Mouse (Mus musculus). Detection Range: 16-1000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P01132. Gene ID: 13645. Target Synonyms: Epidermal growth factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1852