Learn More
Biomatik Corporation IL-10 (Murine) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51894
Encompass_Preferred
IL-10 (Murine) Sandwich ELISA Kit for detecting IL-10 in plasma, serum, lysate. Species Reactivity: Mouse (Mus musculus). Detection Range: 47-3000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P18893. Gene ID: 16153. Target Synonyms: Interleukin-10; Cytokine synthesis inhibitory factor; CSIF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1858