Learn More
Biomatik Corporation IL-17 (Murine) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51897
Encompass_Preferred
IL-17 (Murine) Sandwich ELISA Kit for detecting IL-17 in plasma, serum, lysate. Species Reactivity: Mouse (Mus musculus). Detection Range: 16-1000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q62386. Gene ID: 16171. Target Synonyms: Interleukin-17A; Cytotoxic T-lymphocyte-associated antigen 8; CTLA-8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1861