Learn More
Biomatik Corporation IL-4 (Murine) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51903
Encompass_Preferred
IL-4 (Murine) Sandwich ELISA Kit for detecting IL-4 in plasma, serum, lysate. Species Reactivity: Mouse (Mus musculus). Detection Range: 16-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P07750. Gene ID: 16189. Target Synonyms: Interleukin-4; B-cell IgG differentiation factor; B-cell growth factor 1; B-cell stimulatory factor 1; BSF-1; IGG1 induction factor; Lymphocyte stimulatory factor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1865