Learn More
Biomatik Corporation MDC (Murine) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51910
Encompass_Preferred
MDC (Murine) Sandwich ELISA Kit for detecting MDC in plasma, serum, lysate. Species Reactivity: Mouse (Mus musculus). Detection Range: 8-1000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O88430. Gene ID: 20299. Target Synonyms: C-C motif chemokine 22; Activated B and dendritic cell-derived; CC chemokine ABCD-1; Small-inducible cytokine A22; Abcd1; Scya22. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1867