Learn More
Biomatik Corporation IFN-gamma (Rat) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51921
Encompass_Preferred
IFN-gamma (Rat) Sandwich ELISA Kit for detecting IFN-gamma in plasma, serum, lysate. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 32-4000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P01581. Gene ID: 25712. Target Synonyms: Interferon gamma; IFN-gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1876