Learn More
Biomatik Corporation RANTES (Rat) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51929
Encompass_Preferred
RANTES (Rat) Sandwich ELISA Kit for detecting RANTES in plasma, serum, lysate. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 16-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P50231. Gene ID: 81780. Target Synonyms: C-C motif chemokine 5; SIS-delta; Small-inducible cytokine A5; T-cell-specific protein RANTES; Scya5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1880