Learn More
Biomatik Corporation IGF-1R (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51942
Encompass_Preferred
IGF-1R (Human) Sandwich ELISA Kit for detecting IGF-1R in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 250-16000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P08069. Gene ID: 3480. Target Synonyms: Insulin-like growth factor 1 receptor; CD221. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1888