Learn More
Biomatik Corporation IL-17BR (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51954
Encompass_Preferred
IL-17BR (Human) Sandwich ELISA Kit for detecting IL-17BR in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 157-10000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9NRM6. Gene ID: 55540. Target Synonyms: Interleukin-17 receptor B; Cytokine receptor-like 4; IL-17 receptor homolog 1; CRL4; EVI27; IL17BR. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1898