Learn More
Biomatik Corporation M-CSF (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51967
Encompass_Preferred
M-CSF (Human) Sandwich ELISA Kit for detecting M-CSF in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 16-1000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P09603. Gene ID: 1435. Target Synonyms: Macrophage colony-stimulating factor 1; Lanimostim. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1905