Learn More
Biomatik Corporation OSM-R (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51973
Encompass_Preferred
OSM-R (Human) Sandwich ELISA Kit for detecting OSM-R in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 313-20000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q99650. Gene ID: 9180. Target Synonyms: Oncostatin-M-specific receptor subunit beta; Interleukin-31 receptor subunit beta; OSMRB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1908