Learn More
Biomatik Corporation VEGF-R2 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51976
Encompass_Preferred
VEGF-R2 (Human) Sandwich ELISA Kit for detecting VEGF-R2 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P35968. Gene ID: 3791. Target Synonyms: Vascular endothelial growth factor receptor 2; Fetal liver kinase 1; Kinase insert domain receptor; Protein-tyrosine kinase receptor flk-1; CD309. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1911