Learn More
Biomatik Corporation TGF-alpha (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51978
Encompass_Preferred
TGF-alpha (Human) Sandwich ELISA Kit for detecting TGF-alpha in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 8-500pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P01135. Gene ID: 7039. Target Synonyms: Transforming growth factor alpha; EGF-like TGF; TGF type 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1913