Learn More
Biomatik Corporation Total MMP-7 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51989
Encompass_Preferred
Total MMP-7 (Human) Sandwich ELISA Kit for detecting Total MMP-7 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P09237. Gene ID: 4316. Target Synonyms: Matrilysin; Matrin; Matrix metalloproteinase-7; Pump-1 protease; Uterine metalloproteinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1918