Learn More
Biomatik Corporation VEGF-R1 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51992
Encompass_Preferred
VEGF-R1 (Human) Sandwich ELISA Kit for detecting VEGF-R1 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 157-10000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P17948. Gene ID: 2321. Target Synonyms: Vascular endothelial growth factor receptor 1; Fms-like tyrosine kinase 1; Tyrosine-protein kinase FRT; Tyrosine-protein kinase receptor FLT; Vascular permeability factor receptor; FLT; FRT. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1921