Learn More
Biomatik Corporation IL-17E (Murine) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51997
Encompass_Preferred
IL-17E (Murine) Sandwich ELISA Kit for detecting IL-17E in plasma, serum, lysate. Species Reactivity: Mouse (Mus musculus). Detection Range: 63-4000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q8VHH8. Gene ID: 140806. Target Synonyms: Interleukin 25; IL-25; Interleukin-17E. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1925