Learn More
Biomatik Corporation IL-5 (Murine) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52005
Encompass_Preferred
IL-5 (Murine) Sandwich ELISA Kit for detecting IL-5 in plasma, serum, lysate. Species Reactivity: Mouse (Mus musculus). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P04401. Gene ID: 16191. Target Synonyms: Interleukin-5; B-cell growth factor II; Cytotoxic T-lymphocyte inducer; Eosinophil differentiation factor; T-cell replacing factor; BCGF-II; TRF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1933