Learn More
Biomatik Corporation Angiopoietin-Like 3 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52012
Encompass_Preferred
Angiopoietin-Like 3 (Human) Sandwich ELISA Kit for detecting Angiopoietin-Like 3 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9Y5C1. Gene ID: 27329. Target Synonyms: Angiopoietin-related protein 3; Angiopoietin-5; ANG-5; ANGPT5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1939