Learn More
Biomatik Corporation BMP-7 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52015
Encompass_Preferred
BMP-7 (Human) Sandwich ELISA Kit for detecting BMP-7 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 63-4000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P18075. Gene ID: 655. Target Synonyms: Bone morphogenetic protein 7; Osteogenic protein 1; Eptotermin alfa; OP-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1941