Learn More
Biomatik Corporation TARC (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52018
Encompass_Preferred
TARC (Human) Sandwich ELISA Kit for detecting TARC in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 8-500pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q92583. Gene ID: 6361. Target Synonyms: C-C motif chemokine 17; CC chemokine TARC; Small-inducible cytokine A17; Thymus and activation-regulated chemokine; SCYA17. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1943