Learn More
Biomatik Corporation Eotaxin-2 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52020
Encompass_Preferred
Eotaxin-2 (Human) Sandwich ELISA Kit for detecting Eotaxin-2 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O00175. Gene ID: 6369. Target Synonyms: C-C motif chemokine 24; CK-beta-6; Eosinophil chemotactic protein 2; Eotaxin-2; Myeloid progenitor inhibitory factor 2; Small-inducible cytokine A24; MPIF-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1945