Learn More
Biomatik Corporation MCP-3 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52022
Encompass_Preferred
MCP-3 (Human) Sandwich ELISA Kit for detecting MCP-3 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 8-500pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P80098. Gene ID: 6354. Target Synonyms: C-C motif chemokine 7; Monocyte chemoattractant protein 3; Monocyte chemotactic protein 3; NC28; Small-inducible cytokine A7; SCYA6; SCYA7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1947