Learn More
Biomatik Corporation Tissue Factor (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52027
Encompass_Preferred
Tissue Factor (Human) Sandwich ELISA Kit for detecting Tissue Factor in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 8-500pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P13726. Gene ID: 2152. Target Synonyms: Coagulation factor III; Thromboplastin; CD142. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1951