Learn More
Biomatik Corporation BCA-1 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52033
Encompass_Preferred
BCA-1 (Human) Sandwich ELISA Kit for detecting BCA-1 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 16-1000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O43927. Gene ID: 10563. Target Synonyms: C-X-C motif chemokine 13; B lymphocyte chemoattractant; CXC chemokine BLC; Small-inducible cytokine B13; Angie. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1954