Learn More
Biomatik Corporation FOLR1 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52041
Encompass_Preferred
FOLR1 (Human) Sandwich ELISA Kit for detecting FOLR1 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 94-6000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P15328. Gene ID: 2348. Target Synonyms: Folate receptor alpha; FR-alpha; Adult folate-binding protein; FBP; Folate receptor 1; KB cells FBP; Ovarian tumor-associated antigen MOv18. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1962