Learn More
Biomatik Corporation PSA (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52047
Encompass_Preferred
PSA (Human) Sandwich ELISA Kit for detecting PSA in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 938-60000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P07288. Gene ID: 354. Target Synonyms: Prostate-specific antigen; PSA; Gamma-seminoprotein; Seminin; P-30 antigen; Semenogelase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1965