Learn More
Biomatik Corporation NGAL (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52050
Encompass_Preferred
NGAL (Human) Sandwich ELISA Kit for detecting NGAL in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 79-5000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P80188. Gene ID: 3934. Target Synonyms: Neutrophil gelatinase-associated lipocalin; 25 kDa alpha-2-microglobulin-related subunit of MMP-9; Lipocalin-2; Oncogene 24p3; p25. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1968