Learn More
Biomatik Corporation Pro-Cathepsin B (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52058
Encompass_Preferred
Pro-Cathepsin B (Human) Sandwich ELISA Kit for detecting Pro-Cathepsin B in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 250-16000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P07858. Gene ID: 1508. Target Synonyms: Cathepsin B; APP secretase; APPS; Cathepsin B1; CPSB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1973