Learn More
Biomatik Corporation sTNF-R2 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52066
Encompass_Preferred
sTNF-R2 (Human) Sandwich ELISA Kit for detecting sTNF-R2 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 8-500pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P20333. Gene ID: 7133. Target Synonyms: Tumor necrosis factor receptor superfamily member 1B; Tumor necrosis factor receptor 2; TNF-R2; Tumor necrosis factor receptor type II; TNF-RII; TNFR-II; p75; p80 TNF-alpha receptor; CD120b; INN=Etanercept; TNFBR; TNFR2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1975