Learn More
Biomatik Corporation Tie-2 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52073
Encompass_Preferred
Tie-2 (Human) Sandwich ELISA Kit for detecting Tie-2 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 157-10000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q02763. Gene ID: 7010. Target Synonyms: Angiopoietin-1 receptor; Tunica interna endothelial cell kinase; Tyrosine-protein kinase receptor TEK; Tyrosine-protein kinase receptor TIE-2; hTIE2; p140 TEK; CD202b; TIE2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1979