Learn More
Biomatik Corporation GM-CSF (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51835
Encompass_Preferred
GM-CSF (Human) Sandwich ELISA Kit for detecting GM-CSF in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-1000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P04141. Gene ID: 1437. Target Synonyms: Granulocyte-macrophage colony-stimulating factor; Colony-stimulating factor; CSF; Molgramostin; Sargramostim. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1815