Learn More
Biomatik Corporation IL-31 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51853
Encompass_Preferred
IL-31 (Human) Sandwich ELISA Kit for detecting IL-31 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-4000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q6EBC2. Gene ID: 386653. Target Synonyms: Interleukin-31. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1829