Learn More
Biomatik Corporation I-TAC (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51862
Encompass_Preferred
I-TAC (Human) Sandwich ELISA Kit for detecting I-TAC in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O14625. Gene ID: 6373. Target Synonyms: C-X-C motif chemokine 11; Beta-R1; H174; Interferon gamma-inducible protein 9; Interferon-inducible T-cell alpha chemoattractant; Small-inducible cytokine B11; SCYB11; SCYB9B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1833