Learn More
Biomatik Corporation sRANK Ligand (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51878
Encompass_Preferred
sRANK Ligand (Human) Sandwich ELISA Kit for detecting sRANK Ligand in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 63-4000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9Y6Q6. Gene ID: 8792. Target Synonyms: Tumor necrosis factor receptor superfamily member 11A; Osteoclast differentiation factor receptor; ODFR; Receptor activator of NF-KB; CD265. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1846