Learn More
Biomatik Corporation TIMP-1 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51880
Encompass_Preferred
TIMP-1 (Human) Sandwich ELISA Kit for detecting TIMP-1 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-1000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P01033. Gene ID: 7076. Target Synonyms: Metalloproteinase inhibitor 1; Erythroid-potentiating activity; Fibroblast collagenase inhibitor; Tissue inhibitor of metalloproteinases 1; EPA; CLGI; TIMP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1848