Learn More
Biomatik Corporation TWEAK (Human) Sandwich ELISA Kit
Catalog No. 502291851
Encompass_Preferred
TWEAK (Human) Sandwich ELISA Kit for detecting TWEAK in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 16-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O43508. Gene ID: 8742. Target Synonyms: Tumor necrosis factor ligand superfamily member 12; APO3 ligand; TNF-related weak inducer of apoptosis; APO3L; DR3LG. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-1851 Supplier Biomatik Corporation Supplier No. EKA51886
TWEAK (Human) Sandwich ELISA Kit for detecting TWEAK in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 16-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O43508. Gene ID: 8742. Target Synonyms: Tumor necrosis factor ligand superfamily member 12; APO3 ligand; TNF-related weak inducer of apoptosis; APO3L; DR3LG. Restrictions: For Research Use Only. Not for use in diagnostic procedures.