Learn More
Biomatik Corporation IGF-1 (Murine) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51893
Encompass_Preferred
IGF-1 (Murine) Sandwich ELISA Kit for detecting IGF-1 in plasma, serum, lysate. Species Reactivity: Mouse (Mus musculus). Detection Range: 62-6000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P05017. Gene ID: 16000. Target Synonyms: Insulin-like growth factor I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1857