Learn More
Biomatik Corporation sRANK Ligand (Murine) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51916
Encompass_Preferred
sRANK Ligand (Murine) Sandwich ELISA Kit for detecting sRANK Ligand in plasma, serum, lysate. Species Reactivity: Mouse (Mus musculus). Detection Range: 625-5000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O35305. Gene ID: 21934. Target Synonyms: Tumor necrosis factor receptor superfamily member 11A; Osteoclast differentiation factor receptor; ODFR; Receptor activator of NF-KB; CD265; Rank. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1873