Learn More
Biomatik Corporation GRO/KC (Rat) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51920
Encompass_Preferred
GRO/KC (Rat) Sandwich ELISA Kit for detecting GRO/KC in plasma, serum, lysate. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 16-1000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P14095. Gene ID: 81503. Target Synonyms: Growth-regulated alpha protein; C-X-C motif chemokine 1; Cytokine-induced neutrophil chemoattractant 1; CINC-1; Platelet-derived growth factor-inducible protein KC; Gro; Scyb1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1875