Learn More
Biomatik Corporation SCF (Rat) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51930
Encompass_Preferred
SCF (Rat) Sandwich ELISA Kit for detecting SCF in plasma, serum, lysate. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 16-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P21581. Gene ID: 60427. Target Synonyms: Kit ligand; Hematopoietic growth factor KL; Mast cell growth factor; Stem cell factor; MGF; c-Kit ligand; Kitl; Mgf. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1881