Learn More
Biomatik Corporation VEGF (Rat) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51932
Encompass_Preferred
VEGF (Rat) Sandwich ELISA Kit for detecting VEGF in plasma, serum, lysate. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P16612. Gene ID: 83785. Target Synonyms: Vascular endothelial growth factor A/B; Vascular permeability factor; VEGF-related factor; VRF; VPF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1882