Learn More
Biomatik Corporation Fas Ligand (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51935
Encompass_Preferred
Fas Ligand (Human) Sandwich ELISA Kit for detecting Fas Ligand in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P48023. Gene ID: 356. Target Synonyms: Tumor necrosis factor ligand superfamily member 6; Apoptosis antigen ligand; CD95 ligand; Fas antigen ligand; CD178; APT1LG1; CD95L; FASL; TNFSF6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1883