Learn More
Biomatik Corporation Endoglin (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51975
Encompass_Preferred
Endoglin (Human) Sandwich ELISA Kit for detecting Endoglin in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P17813. Gene ID: 2022. Target Synonyms: Endoglin; CD105; END. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1910