Learn More
Biomatik Corporation TIMP-2 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51983
Encompass_Preferred
TIMP-2 (Human) Sandwich ELISA Kit for detecting TIMP-2 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P16035. Gene ID: 7077. Target Synonyms: Metalloproteinase inhibitor 2; CSC-21K; Tissue inhibitor of metalloproteinases 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1914